Assay Details
| Gene Symbol: LOC107049165 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_025142807.1 | XP_024998575.1 | 17 - 18 | 2307 | 115 | |
| GenBank mRNA | JN248386.1 | - | 9 - 10 | 1138 | 115 | |
More Information
Set Membership: |
|

LOC107049165
Genomic Map