Assay Details
| Gene Symbol: LOC107051537 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_025145500.1 | XP_025001268.1 | 1 - 2 | 267 | 105 | |
| GenBank mRNA | X73973.2 | - | 345 | 104 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_025145500.1 transcripts.
Set Membership: |
|

LOC107051537
Genomic Map