Assay Details
| Gene Symbol: SLC25A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_025155588.1 | XP_025011356.1 | 2 - 3 | 1119 | 54 | |
| XM_415059.6 | XP_415059.2 | 2 - 3 | 270 | 54 | ||
| GenBank mRNA | BX932527.1 | - | 2 - 3 | 221 | 54 | |
| BX933737.2 | - | 2 - 3 | 212 | 54 | ||
More Information
Set Membership: |
|

SLC25A1
Genomic Map