Assay Details
| Gene Symbol: ACKR4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_025147497.1 | XP_025003265.1 | 3 | 762 | 63 | |
| GenBank mRNA | BX932809.2 | - | 1 | 216 | 63 | |
| BX950650.1 | - | 2 | 335 | 63 | ||
| BX950657.1 | - | 2 | 334 | 63 | ||
More Information
Set Membership: |
|

ACKR4
Genomic Map