Assay Details
| Gene Symbol: PMVK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015298512.2 | XP_015153998.1 | 4 - 5 | 450 | 63 | |
| XM_025143468.1 | XP_024999236.1 | 3 - 4 | 367 | 63 | ||
| GenBank mRNA | BX932950.1 | - | 4 - 5 | 426 | 63 | |
| BX933122.2 | - | 4 - 5 | 434 | 63 | ||
| BX950466.1 | - | 4 - 5 | 398 | 63 | ||
More Information
Set Membership: |
|

PMVK
Genomic Map