Assay Details
| Gene Symbol: PLCXD3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003642932.4 | XP_003642980.1 | 2 - 3 | 891 | 66 | |
| GenBank mRNA | BX933157.1 | - | 2 - 3 | 864 | 66 | |
More Information
Set Membership: |
|

PLCXD3
Genomic Map