Assay Details
| Gene Symbol: SNRPD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_025144523.1 | XP_025000291.1 | 1 - 2 | 152 | 61 | |
| GenBank mRNA | BX933865.1 | - | 1 - 2 | 63 | 61 | |
| BX936185.1 | - | 1 - 2 | 82 | 61 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_025144523.1 transcripts.
Set Membership: |
|

SNRPD2
Genomic Map