Assay Details
| Gene Symbol: RAD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004937180.3 | XP_004937237.1 | 4 - 5 | 770 | 93 | |
| GenBank mRNA | BX933947.2 | - | 4 - 5 | 683 | 93 | |
| BX935869.2 | - | 4 - 5 | 667 | 93 | ||
More Information
Set Membership: |
|

RAD1
Genomic Map