Assay Details
| Gene Symbol: PIN4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_420136.6 | XP_420136.2 | 3 - 4 | 276 | 78 | |
| GenBank mRNA | BX934304.2 | - | 2 - 3 | 183 | 78 | |
| BX934788.1 | - | 3 - 4 | 248 | 78 | ||
More Information
Set Membership: |
|

PIN4
Genomic Map