Assay Details
| Gene Symbol: NIF3L1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004942604.3 | XP_004942661.1 | 2 - 3 | 691 | 67 | |
| XM_421932.6 | XP_421932.3 | 2 - 3 | 669 | 67 | ||
| GenBank mRNA | BX934332.2 | - | 2 - 3 | 597 | 67 | |
| CR523521.1 | - | 2 - 3 | 608 | 67 | ||
More Information
Set Membership: |
|

NIF3L1
Genomic Map