Assay Details
| Gene Symbol: LOC107051857 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015277055.2 | XP_015132541.2 | 2 - 3 | 1019 | 84 | |
| GenBank mRNA | BX934415.1 | - | 1 - 2 | 131 | 84 | |
| BX950311.2 | - | 173 | 84 | |||
More Information
Set Membership: |
|

LOC107051857
Genomic Map