Assay Details
| Gene Symbol: HTRA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015288895.2 | XP_015144381.1 | 4 - 5 | 1099 | 112 | |
| GenBank mRNA | BX934487.1 | - | 2 - 3 | 230 | 112 | |
| BX950476.2 | - | 4 - 5 | 654 | 112 | ||
More Information
Set Membership: |
|

HTRA1
Genomic Map