Assay Details
| Gene Symbol: ZNHIT3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015296154.2 | XP_015151640.1 | 3 - 4 | 274 | 78 | |
| XM_025142040.1 | XP_024997808.1 | 3 - 4 | 278 | 78 | ||
| XM_025142041.1 | XP_024997809.1 | 4 - 5 | 2481 | 78 | ||
| XM_025142042.1 | XP_024997810.1 | 3 - 4 | 397 | 78 | ||
| GenBank mRNA | BX934721.1 | - | 2 - 3 | 315 | 78 | |
| CR386289.1 | - | 3 - 4 | 270 | 78 | ||
More Information
Set Membership: |
|

ZNHIT3
Genomic Map