Assay Details
| Gene Symbol: WFDC8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015296579.1 | XP_015152065.1 | 12 - 13 | 2269 | 91 | |
| XM_015296580.1 | XP_015152066.1 | 11 - 12 | 2125 | 91 | ||
| XM_015296581.2 | XP_015152067.1 | 13 - 14 | 2339 | 91 | ||
| XM_015296582.2 | XP_015152068.1 | 12 - 13 | 2122 | 91 | ||
| GenBank mRNA | BX934784.2 | - | 3 - 4 | 476 | 91 | |
More Information
Set Membership: |
|

WFDC8
Genomic Map