Assay Details
| Gene Symbol: CLIC3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003642249.4 | XP_003642297.3 | 5 - 6 | 667 | 58 | |
| GenBank mRNA | BX935133.2 | - | 5 - 6 | 549 | 58 | |
More Information
Set Membership: |
|

CLIC3
Genomic Map