Assay Details
| Gene Symbol: BYSL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_418047.6 | XP_418047.3 | 715 | 64 | ||
| GenBank mRNA | BX935205.2 | - | 341 | 64 | ||
| CR524293.1 | - | 344 | 64 | |||
More Information
Set Membership: |
|

BYSL
Genomic Map