Assay Details
| Gene Symbol: MAMDC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003643015.3 | XP_003643063.1 | 6 - 7 | 1021 | 82 | |
| GenBank mRNA | BX935251.1 | - | 6 - 7 | 1007 | 82 | |
More Information
Set Membership: |
|

MAMDC2
Genomic Map