Assay Details
| Gene Symbol: PPIC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001231332.5 | XP_001231333.3 | 1 - 2 | 1727 | 121 | |
| GenBank mRNA | BX935257.1 | - | 1 - 2 | 108 | 121 | |
| CR352486.1 | - | 1 - 2 | 179 | 121 | ||
More Information
Set Membership: |
|

PPIC
Genomic Map