Assay Details
| Gene Symbol: FSD1L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004949352.3 | XP_004949409.1 | 6 - 7 | 538 | 77 | |
| XM_004949353.3 | XP_004949410.1 | 6 - 7 | 538 | 77 | ||
| XM_004949354.3 | XP_004949411.1 | 6 - 7 | 538 | 77 | ||
| XM_424884.6 | XP_424884.2 | 6 - 7 | 538 | 77 | ||
| GenBank mRNA | BX935589.2 | - | 6 - 7 | 538 | 77 | |
| CR389170.1 | - | 5 - 6 | 479 | 77 | ||
More Information
Set Membership: |
|

FSD1L
Genomic Map