Assay Details
| Gene Symbol: MED29 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015300221.1 | XP_015155707.1 | 1 - 2 | 244 | 73 | |
| GenBank mRNA | BX935654.1 | - | 1 - 2 | 260 | 73 | |
More Information
Set Membership: |
|

MED29
Genomic Map