Assay Details
| Gene Symbol: PRDX4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001233999.3 | XP_001234000.1 | 3 - 4 | 479 | 67 | |
| XM_416800.6 | XP_416800.2 | 3 - 4 | 657 | 67 | ||
| GenBank mRNA | BX935739.1 | - | 3 - 4 | 488 | 67 | |
More Information
Set Membership: |
|

PRDX4
Genomic Map