Assay Details
| Gene Symbol: FAM19A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004944676.3 | XP_004944733.2 | 2 - 3 | 694 | 73 | |
| XM_015293256.2 | XP_015148742.1 | 2 - 3 | 150 | 73 | ||
| GenBank mRNA | BX936093.2 | - | 2 - 3 | 145 | 73 | |
More Information
Set Membership: |
|

FAM19A1
Genomic Map