Assay Details
| Gene Symbol: LOC101749201 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015297728.2 | XP_015153214.2 | 2 - 3 | 194 | 82 | |
| GenBank mRNA | BX936149.2 | - | 1 - 2 | 295 | 82 | |
More Information
Set Membership: |
|

LOC101749201
Genomic Map