Assay Details
| Gene Symbol: DNAJB4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004936669.3 | XP_004936726.1 | 4 | 78 | ||
| XM_422386.6 | XP_422386.1 | 5 | 78 | |||
| GenBank mRNA | BX950520.1 | - | 1 | 120 | 78 | |
More Information
Set Membership: |
|

DNAJB4
Genomic Map