Assay Details
| Gene Symbol: TDH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_420039.6 | XP_420039.1 | 6 - 7 | 654 | 85 | |
| GenBank mRNA | BX950580.1 | - | 5 - 6 | 641 | 85 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

TDH| Gene Symbol: TDH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_420039.6 | XP_420039.1 | 6 - 7 | 654 | 85 | |
| GenBank mRNA | BX950580.1 | - | 5 - 6 | 641 | 85 | |
Set Membership: |
|