Assay Details
| Gene Symbol: DERL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015295984.2 | XP_015151470.1 | 6 - 7 | 633 | 116 | |
| GenBank mRNA | BX950582.2 | - | 2 - 3 | 858 | 116 | |
More Information
Set Membership: |
|

DERL2
Genomic Map