Assay Details
| Gene Symbol: AVPI1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015288797.2 | XP_015144283.1 | 2 - 3 | 443 | 63 | |
| GenBank mRNA | BX950736.1 | - | 1 - 2 | 631 | 63 | |
More Information
Set Membership: |
|

AVPI1
Genomic Map