Assay Details
| Gene Symbol: LOC100859602 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_025144610.1 | XP_025000378.1 | 11 - 12 | 2440 | 74 | |
| XM_025144611.1 | XP_025000379.1 | 12 - 13 | 2051 | 74 | ||
More Information
Set Membership: |
|

LOC100859602
Genomic Map