Assay Details
| Gene Symbol: LRR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_025151274.1 | XP_025007042.1 | 2 | 1644 | 95 | |
| XM_421455.6 | XP_421455.3 | 3 | 1743 | 95 | ||
| GenBank mRNA | CR386704.1 | - | 1 | 420 | 95 | |
More Information
Set Membership: |
|

LRR1
Genomic Map