Assay Details
| Gene Symbol: CHST14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003641286.4 | XP_003641334.3 | 1 | 2205 | 85 | |
| GenBank mRNA | CR387702.1 | - | 2 | 1015 | 85 | |
More Information
Set Membership: |
|

CHST14
Genomic Map