Assay Details
| Gene Symbol: TMEM252 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003642915.3 | XP_003642963.2 | 2 | 901 | 102 | |
| GenBank mRNA | CR388546.1 | - | 2 | 614 | 102 | |
More Information
Set Membership: |
|

TMEM252
Genomic Map