Assay Details
| Gene Symbol: DKK4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015278709.2 | XP_015134195.1 | 4 | 1107 | 118 | |
| GenBank mRNA | AY049017.1 | - | 2 | 520 | 118 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

DKK4| Gene Symbol: DKK4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015278709.2 | XP_015134195.1 | 4 | 1107 | 118 | |
| GenBank mRNA | AY049017.1 | - | 2 | 520 | 118 | |
Set Membership: |
|