Assay Details
| Gene Symbol: SLC3A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004935370.3 | XP_004935427.1 | 8 | 1431 | 91 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

SLC3A1| Gene Symbol: SLC3A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004935370.3 | XP_004935427.1 | 8 | 1431 | 91 | |
Set Membership: |
|