Assay Details
| Gene Symbol: IDH1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004942682.3 | XP_004942739.1 | 9 | 1473 | 65 | |
| XM_015289549.2 | XP_015145035.1 | 9 | 1578 | 65 | ||
| XM_015289550.2 | XP_015145036.1 | 10 | 1482 | 65 | ||
| XM_025152402.1 | XP_025008170.1 | 10 | 1553 | 65 | ||
| XM_025152403.1 | XP_025008171.1 | 10 | 1586 | 65 | ||
More Information
Set Membership: |
|

IDH1
Genomic Map