Assay Details
| Gene Symbol: UTS2R | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004946094.3 | XP_004946151.1 | 2 | 2007 | 103 | |
| XM_025141679.1 | XP_024997447.1 | 2 | 1021 | 103 | ||
| GenBank mRNA | KU887758.1 | - | 2 | 1178 | 103 | |
More Information
Set Membership: |
|

UTS2R
Genomic Map