Assay Details
| Gene Symbol: UGT8L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_426504.6 | XP_426504.2 | 6 | 1598 | 93 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

UGT8L| Gene Symbol: UGT8L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_426504.6 | XP_426504.2 | 6 | 1598 | 93 | |
Set Membership: |
|