Assay Details
| Gene Symbol: CNGA4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015280937.2 | XP_015136423.1 | 5 | 2285 | 91 | |
| XM_015280938.2 | XP_015136424.1 | 5 | 2263 | 91 | ||
| XM_025146967.1 | XP_025002735.1 | 4 | 2650 | 91 | ||
More Information
Set Membership: |
|

CNGA4
Genomic Map