Assay Details
| Gene Symbol: PIGZ | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_025153617.1 | XP_025009385.1 | 2 | 680 | 62 | |
| XM_426699.5 | XP_426699.2 | 2 | 646 | 62 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

PIGZ| Gene Symbol: PIGZ | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_025153617.1 | XP_025009385.1 | 2 | 680 | 62 | |
| XM_426699.5 | XP_426699.2 | 2 | 646 | 62 | ||
Set Membership: |
|