Assay Details
| Gene Symbol: MRPS28 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001233657.4 | XP_001233658.3 | 3 | 1014 | 118 | |
| GenBank mRNA | BX931469.2 | - | 3 | 622 | 118 | |
| BX932153.2 | - | 3 | 637 | 118 | ||
| BX932347.1 | - | 3 | 697 | 118 | ||
| BX935848.2 | - | 3 | 611 | 118 | ||
More Information
Set Membership: |
|

MRPS28
Genomic Map