Assay Details
| Gene Symbol: ATP5E | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BX931735.1 | - | 2 | 325 | 119 | |
| BX933912.1 | - | 2 | 312 | 119 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ATP5E| Gene Symbol: ATP5E | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BX931735.1 | - | 2 | 325 | 119 | |
| BX933912.1 | - | 2 | 312 | 119 | ||
Set Membership: |
|