Assay Details
| Gene Symbol: GPR141 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004939529.3 | XP_004939586.1 | 2 | 938 | 93 | |
| XM_015281687.2 | XP_015137173.1 | 2 | 1102 | 93 | ||
| GenBank mRNA | KX928073.1 | - | 2 | 914 | 93 | |
| KX928074.1 | - | 2 | 1060 | 93 | ||
| KX928075.1 | - | 3 | 1332 | 93 | ||
| KX928076.1 | - | 3 | 1478 | 93 | ||
More Information
Set Membership: |
|

GPR141
Genomic Map