Assay Details
| Gene Symbol: LIPT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015281019.2 | XP_015136505.1 | 1 | 306 | 168 | |
| GenBank mRNA | BX933412.2 | - | 1 | 226 | 168 | |
| BX933916.2 | - | 1 | 245 | 168 | ||
| BX950671.2 | - | 1 | 298 | 168 | ||
More Information
Set Membership: |
|

LIPT2
Genomic Map