Assay Details
| Gene Symbol: PROK2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015293233.2 | XP_015148719.1 | 3 | 5194 | 87 | |
| GenBank mRNA | BX933454.1 | - | 3 | 223 | 87 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

PROK2| Gene Symbol: PROK2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015293233.2 | XP_015148719.1 | 3 | 5194 | 87 | |
| GenBank mRNA | BX933454.1 | - | 3 | 223 | 87 | |
Set Membership: |
|