Assay Details
| Gene Symbol: LOC107050443 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_025145750.1 | XP_025001518.1 | 7 - 8 | 572 | 60 | |
| XM_025145751.1 | XP_025001519.1 | 7 - 8 | 571 | 60 | ||
More Information
Set Membership: |
|

LOC107050443
Genomic Map