Assay Details
| Gene Symbol: CIART | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015298641.2 | XP_015154127.2 | 1 - 2 | 191 | 122 | |
| XM_025143657.1 | XP_024999425.1 | 1 - 2 | 605 | 122 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015298641.2 transcripts.
Set Membership: |
|

CIART
Genomic Map