Assay Details
| Gene Symbol: TMEM98 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015299412.2 | XP_015154898.1 | 4 - 5 | 529 | 64 | |
| XM_015299413.2 | XP_015154899.1 | 4 - 5 | 542 | 64 | ||
| XM_015299415.2 | XP_015154901.1 | 4 - 5 | 467 | 64 | ||
| XM_025144006.1 | XP_024999774.1 | 4 - 5 | 543 | 64 | ||
More Information
Set Membership: |
|

TMEM98
Genomic Map