Assay Details
| Gene Symbol: C4H4ORF45 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001233137.5 | XP_001233138.5 | 2 - 3 | 1430 | 79 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

C4H4ORF45| Gene Symbol: C4H4ORF45 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001233137.5 | XP_001233138.5 | 2 - 3 | 1430 | 79 | |
Set Membership: |
|