Assay Details
| Gene Symbol: FAM126B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004942607.3 | XP_004942664.1 | 10 - 11 | 1001 | 66 | |
| XM_015289464.2 | XP_015144950.1 | 11 - 12 | 1082 | 66 | ||
| XM_015289465.2 | XP_015144951.1 | 12 - 13 | 1265 | 66 | ||
| XM_015289466.2 | XP_015144952.1 | 11 - 12 | 1086 | 66 | ||
| XM_015289467.2 | XP_015144953.1 | 12 - 13 | 1284 | 66 | ||
| XM_015289468.2 | XP_015144954.1 | 11 - 12 | 1253 | 66 | ||
| XM_015289469.2 | XP_015144955.1 | 10 - 11 | 1297 | 66 | ||
| XM_015289470.2 | XP_015144956.1 | 11 - 12 | 1082 | 66 | ||
More Information
Set Membership: |
|

FAM126B
Genomic Map