Assay Details
| Gene Symbol: PAPD5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015292408.2 | XP_015147894.1 | 11 - 12 | 2027 | 62 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

PAPD5| Gene Symbol: PAPD5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015292408.2 | XP_015147894.1 | 11 - 12 | 2027 | 62 | |
Set Membership: |
|