Assay Details
| Gene Symbol: GPR61 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015298772.2 | XP_015154258.2 | 303 | 66 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015298772.2 transcripts.
Set Membership: |
|

GPR61
Genomic Map